Learn More
Biomatik Corporation Mouse Alkaline Phosphatase, Liver/Bone/Kidney (ALPL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02247
Encompass_Preferred
Mouse Alkaline Phosphatase, Tissue-nonspecific (ALPL) ELISA Kit for detecting ALPL in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P09242. Gene ID: 11647. Target Synonyms: BALP; BSAP; HOPS; AP-TNAP; TNSALP; Bone Alkaline Phosphatase; Alkaline Phosphatase,Tissue-Nonspecific Isozyme; Alkaline Phosphatase, Liver/Bone/Kidney. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1740