Learn More
Biomatik Corporation Rat Alkaline Phosphatase, Liver/Bone/Kidney (ALPL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02248
Encompass_Preferred
Rat Alkaline Phosphatase, Tissue-nonspecific (ALPL) ELISA Kit for detecting ALPL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.15ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08289. Gene ID: 25586. Target Synonyms: BALP; BSAP; HOPS; AP-TNAP; TNSALP; Bone Alkaline Phosphatase; Alkaline Phosphatase,Tissue-Nonspecific Isozyme; Alkaline Phosphatase, Liver/Bone/Kidney. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1741