Learn More
Biomatik Corporation Human Allograft Inflammatory Factor 1 (AIF1) ELISA Kit, 96 tests
Catalog No. 501501744
Encompass_Preferred
The Human Allograft Inflammatory Factor 1 (AIF1) ELISA Kit allows for quantitative determination of Allograft Inflammatory Factor 1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2,000 pg/mL. Sensitivity: 13.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P55008. Cross Activity: No significant cross-reactivity or interference between Allograft Inflammatory Factor 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1744 Supplier Biomatik Corporation Supplier No. EKU02254
The Human Allograft Inflammatory Factor 1 (AIF1) ELISA Kit allows for quantitative determination of Allograft Inflammatory Factor 1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2,000 pg/mL. Sensitivity: 13.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P55008. Cross Activity: No significant cross-reactivity or interference between Allograft Inflammatory Factor 1 and analogues was observed. Restriction: For Research Use Only.