Learn More
Biomatik Corporation Mouse Allograft Inflammatory Factor 1 (AIF1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02255
Encompass_Preferred
Mouse Ionized Calcium-binding Adapter Molecule 1 (IBA1) ELISA Kit for detecting IBA1 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O70200. Gene ID: 11629. Target Synonyms: AIF1; IRT1; Daintain; Protein G1; Allograft Inflammatory Factor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1745