Learn More
Biomatik Corporation Cattle Alpha-1-Acid Glycoprotein (a1AGP) ELISA Kit, 96 tests
Catalog No. 501501746
Encompass_Preferred
The Cattle Alpha-1-Acid Glycoprotein (a1AGP) ELISA Kit allows for quantitative determination of Alpha-1-Acid Glycoprotein in Serum, Plasma, Urine and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 15.6-1,000 ng/mL. Sensitivity: 7.0 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q5GN72. Cross Activity: No significant cross-reactivity or interference between Alpha-1-Acid Glycoprotein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1746 Supplier Biomatik Corporation Supplier No. EKU02259
The Cattle Alpha-1-Acid Glycoprotein (a1AGP) ELISA Kit allows for quantitative determination of Alpha-1-Acid Glycoprotein in Serum, Plasma, Urine and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 15.6-1,000 ng/mL. Sensitivity: 7.0 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q5GN72. Cross Activity: No significant cross-reactivity or interference between Alpha-1-Acid Glycoprotein and analogues was observed. Restriction: For Research Use Only.