Learn More
Biomatik Corporation Mouse Alpha-1-Acid Glycoprotein (a1AGP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02261
Encompass_Preferred
Mouse Alpha-1-Acid Glycoprotein (a1AGP) ELISA Kit for detecting a1AGP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4,000ng/ml. Sensitivity: 24.5ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q60590. Gene ID: 18405. Target Synonyms: AGP1; a1-AGP; ORM1; OMD 1; Orosomucoid 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1748