Learn More
Biomatik Corporation Rat Alpha-1-Antitrypsin (a1AT) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02266
Encompass_Preferred
Rat Alpha-1-Antitrypsin (a1AT) ELISA Kit for detecting a1AT in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.8-500ng/ml. Sensitivity: 3.5ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P17475. Gene ID: 24648. Target Synonyms: SERPINA1; SPAAT; A1-AT; PI; Serpin Peptidase Inhibitor,Clade A (Alpha-1 Antiproteinase/AntiTrypsin) Member 1; Alpha-1 protease inhibitor; Short peptide from AAT; Serpin A1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1752