Learn More
Biomatik Corporation Cattle Alpha-2-Heremans Schmid Glycoprotein (aHSG) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02273
Encompass_Preferred
The Cattle Alpha-2-Heremans Schmid Glycoprotein (aHSG) ELISA Kit allows for quantitative determination of Alpha-2-Heremans Schmid Glycoprotein in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 6.25-400 ng/mL. Sensitivity: 2.45 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P12763. Cross Activity: No significant cross-reactivity or interference between Alpha-2-Heremans Schmid Glycoprotein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1755