Learn More
Biomatik Corporation Mouse Alpha-2-Heremans Schmid Glycoprotein (aHSG) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02275
Encompass_Preferred
Mouse Alpha-2-Heremans Schmid Glycoprotein (AHSG) ELISA Kit for detecting AHSG in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.71ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P29699. Gene ID: 11625. Target Synonyms: aHSG; HSGa; A2HS; AHS; FETUA; HSG-A2; Fetuin-A; Alpha 2-HS Glycoprotein; Fetuin A; Alpha-2-Z-Globulin; Ba-alpha-2-Glycoprotein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1757