Learn More
Biomatik Corporation Human Alpha-2-Macroglobulin (a2M) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02278
Encompass_Preferred
The Human Alpha-2-Macroglobulin (a2M) ELISA Kit allows for quantitative determination of Alpha-2-Macroglobulin in Serum, Plasma, Urine and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.35 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01023. Cross Activity: No significant cross-reactivity or interference between Alpha-2-Macroglobulin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1759