Learn More
Biomatik Corporation Pig Alpha-Fetoprotein (aFP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02293
Encompass_Preferred
Pig Alpha-Fetoprotein (AFP) ELISA Kit for detecting AFP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.312-20ng/ml. Sensitivity: 0.122ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8MJ76. Gene ID: 397586. Target Synonyms: aFP; A-FP; FETA; HPAFP; Alpha-Fetoglobulin; Alpha-1-fetoprotein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1764