Learn More
Biomatik Corporation Mouse Amine Oxidase, Copper Containing 3 (AOC3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02308
Encompass_Preferred
Mouse Amine Oxidase, Copper Containing 3 (AOC3) ELISA Kit for detecting AOC3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.65ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O70423. Gene ID: 11754. Target Synonyms: VAP1; HPAO; SSAO; Membrane primary amine oxidase; Vascular Adhesion Protein 1; Semicarbazide-Sensitive Amine Oxidase; Copper-Containing Amine Oxygen Oxidoreductase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1768