Learn More
Biomatik Corporation Human Aminoacyl tRNA Synthetase Complex Interacting Multifunctional Protein 1 (AIMP1) ELISA Kit, 96 tests
Catalog No. 501501769
Encompass_Preferred
The Human Aminoacyl tRNA Synthetase Complex Interacting Multifunctional Protein 1 (AIMP1) ELISA Kit allows for quantitative determination of Aminoacyl tRNA Synthetase Complex Interacting Multifunctional Protein 1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 125-8000 pg/mL. Sensitivity: 44 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q12904. Cross Activity: No significant cross-reactivity or interference between Aminoacyl tRNA Synthetase Complex Interacting Multifunctional Protein 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1769 Supplier Biomatik Corporation Supplier No. EKU02309
The Human Aminoacyl tRNA Synthetase Complex Interacting Multifunctional Protein 1 (AIMP1) ELISA Kit allows for quantitative determination of Aminoacyl tRNA Synthetase Complex Interacting Multifunctional Protein 1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 125-8000 pg/mL. Sensitivity: 44 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q12904. Cross Activity: No significant cross-reactivity or interference between Aminoacyl tRNA Synthetase Complex Interacting Multifunctional Protein 1 and analogues was observed. Restriction: For Research Use Only.