Learn More
Biomatik Corporation Rat Amphiregulin (AREG) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02321
Encompass_Preferred
Rat Amphiregulin (AREG) ELISA Kit for detecting AREG in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 11.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P24338. Gene ID: 29183. Target Synonyms: AR; CRDGF; SDGF; Colorectum Cell-Derived Growth Factor; Schwannoma-Derived Growth Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1775