Learn More
Biomatik Corporation Human Amylase Alpha 1, Salivary (AMY1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02323
Encompass_Preferred
The Human Amylase Alpha 1, Salivary (AMY1) ELISA Kit allows for quantitative determination of Amylase Alpha 1, Salivary in Serum, Plasma, Saliva and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.22 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P04745. Cross Activity: No significant cross-reactivity or interference between Amylase Alpha 1, Salivary and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1776