Learn More
Biomatik Corporation Mouse Amyloid Beta Peptide 1-42 (Ab1-42) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02332
Encompass_Preferred
Mouse Amyloid Beta Peptide 1-42 (Ab1-42) ELISA Kit for detecting Ab1-42 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.70-300pg/ml. Sensitivity: 1.36pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P12023. Gene ID: 11820. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1780