Learn More
Biomatik Corporation Rat Androgen Receptor (AR) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02344
Encompass_Preferred
Rat Androgen Receptor (AR) ELISA Kit for detecting AR in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P15207. Gene ID: 24208. Target Synonyms: AIS; DHTR; HUMARA; KD; NR3C4; SBMA; SMAX1; TFM; Dihydrotestosterone receptor; Nuclear Receptor Subfamily 3,Group C,Member 4; Testicular Feminization; Kennedy Disease. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1784