Learn More
Biomatik Corporation Human Angiopoietin 1 (ANGPT1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02358
Encompass_Preferred
The Human Angiopoietin 1 (ANGPT1) ELISA Kit allows for quantitative determination of Angiopoietin 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.24-15 ng/mL. Sensitivity: 0.11 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q15389. Cross Activity: No significant cross-reactivity or interference between Angiopoietin 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1787