Learn More
Biomatik Corporation Rat Angiopoietin 1 (ANGPT1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02360
Encompass_Preferred
Rat Angiopoietin 1 (ANGPT1) ELISA Kit for detecting ANGPT1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O35460. Gene ID: 89807. Target Synonyms: ANG1; AGP1; AGPT; ANG-I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1789