Learn More
Biomatik Corporation Rat Angiopoietin Like Protein 4 (ANGPTL4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02376
Encompass_Preferred
Rat Angiopoietin Like Protein 4 (ANGPTL4) ELISA Kit for detecting ANGPTL4 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.57ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6TMA8. Gene ID: 362850. Target Synonyms: ARP4; FIAF; HFARP; NL2; PGAR; Pp1158; Fasting-Induced Adipose Factor; Hepatic Angiopoietin-Related Protein; Hepatic Fibrinogen/Angiopoietin-Related Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1796