Learn More
Biomatik Corporation Mouse Angiopoietin Like Protein 8 (ANGPTL8) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02381
Encompass_Preferred
Mouse Angiopoietin Like Protein 8 (ANGPTL8) ELISA Kit for detecting ANGPTL8 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8R1L8. Gene ID: 624219. Target Synonyms: RIFL; TD26; C19orf80; Lipasin; Betatrophin; Hepatocellular Carcinoma-Associated Protein TD26. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1798