Learn More
Biomatik Corporation Mouse Angiostatin (ANG) ELISA Kit, 96 tests
Catalog No. 501501800
Encompass_Preferred
Mouse Angiostatin (ANG) ELISA Kit for detecting ANG in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.134ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P20918. Gene ID: 18815. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1800 Supplier Biomatik Corporation Supplier No. EKU02383
Mouse Angiostatin (ANG) ELISA Kit for detecting ANG in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.134ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P20918. Gene ID: 18815. Restrictions: For Research Use Only. Not for use in diagnostic procedures.