Learn More
Biomatik Corporation Human Angiotensin I Converting Enzyme 2 (ACE2) ELISA Kit, 96 tests
Catalog No. 501501804
Encompass_Preferred
The Human Angiotensin I Converting Enzyme 2 (ACE2) ELISA Kit allows for quantitative determination of Angiotensin I Converting Enzyme 2 in Serum, Plasma, and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9BYF1. Cross Activity: No significant cross-reactivity or interference between Angiotensin I Converting Enzyme 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1804 Supplier Biomatik Corporation Supplier No. EKU02399
The Human Angiotensin I Converting Enzyme 2 (ACE2) ELISA Kit allows for quantitative determination of Angiotensin I Converting Enzyme 2 in Serum, Plasma, and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9BYF1. Cross Activity: No significant cross-reactivity or interference between Angiotensin I Converting Enzyme 2 and analogues was observed. Restriction: For Research Use Only.