Learn More
Biomatik Corporation Mouse Angiotensin I Converting Enzyme 2 (ACE2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02400
Encompass_Preferred
Mouse Angiotensin I Converting Enzyme 2 (ACE2) ELISA Kit for detecting ACE2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4,000pg/ml. Sensitivity: 26.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8R0I0. Gene ID: 70008. Target Synonyms: ACEH; ACEII; ACE-II; Peptidyl-Dipeptidase A; ACE-related carboxypeptidase; Angiotensin-converting enzyme homolog; Metalloprotease MPROT15. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1805