Learn More
Biomatik Corporation Rat Angiotensin I Converting Enzyme 2 (ACE2) ELISA Kit, 96 tests
Catalog No. 501501806
Encompass_Preferred
Rat Angiotensin I Converting Enzyme 2 (ACE2) ELISA Kit for detecting ACE2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5EGZ1. Gene ID: 302668. Target Synonyms: ACEH; ACEII; ACE-II; Peptidyl-Dipeptidase A; ACE-related carboxypeptidase; Angiotensin-converting enzyme homolog; Metalloprotease MPROT15. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1806 Supplier Biomatik Corporation Supplier No. EKU02401
Rat Angiotensin I Converting Enzyme 2 (ACE2) ELISA Kit for detecting ACE2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5EGZ1. Gene ID: 302668. Target Synonyms: ACEH; ACEII; ACE-II; Peptidyl-Dipeptidase A; ACE-related carboxypeptidase; Angiotensin-converting enzyme homolog; Metalloprotease MPROT15. Restrictions: For Research Use Only. Not for use in diagnostic procedures.