Learn More
Biomatik Corporation Rat Angiotensin II (AngII) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02407
Encompass_Preferred
Rat Angiotensin II (AngII) ELISA Kit for detecting AngII in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 24.69-2,000pg/ml. Sensitivity: 9.17pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01015. Gene ID: 24179. Target Synonyms: Ang-II; Angiotensin-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1809