Learn More
Biomatik Corporation Rat Angiotensin II Receptor 1 (AGTR1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02410
Encompass_Preferred
Rat Angiotensin II Receptor 1 (AGTR1) ELISA Kit for detecting AGTR1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P25095. Gene ID: 24180. Target Synonyms: AT1R; AG2S; AG2-S; AGTR1A; AGTR1B; AT1; AT1B; AT2R1; AT2-R1; AT2R1A; AT2R1B; HAT1R; AT1 Receptor; Angiotensin Receptor 1B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1811