Learn More
Biomatik Corporation Mouse Angiotensinogen (AGT) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02415
Encompass_Preferred
Mouse Angiotensinogen (AGT) ELISA Kit for detecting AGT in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500ng/ml. Sensitivity: 2.7ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11859. Target Synonyms: ANHU; SERPINA8; Angiotensin; Renin Substrate; Serpin Peptidase Inhibitor,Clade A,Member 8 (Alpha-1 Antiproteinase,Antitrypsin). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1814