Learn More
Biomatik Corporation Rat Angiotensinogen (AGT) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02416
Encompass_Preferred
Rat Angiotensinogen (AGT) ELISA Kit for detecting AGT in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.35ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01015. Gene ID: 24179. Target Synonyms: ANHU; SERPINA8; Angiotensin; Renin Substrate; Serpin Peptidase Inhibitor,Clade A,Member 8 (Alpha-1 Antiproteinase,Antitrypsin). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1815