Learn More
Biomatik Corporation Mouse Annexin A1 (ANXA1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02423
Encompass_Preferred
Mouse Annexin A1 (ANXA1) ELISA Kit for detecting ANXA1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10107. Gene ID: 16952. Target Synonyms: ANX-A1; ANX1; LPC1; Lipocortin I; Chromobindin-9; Calpactin II; Phospholipase A2 inhibitory protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1817