Learn More
Biomatik Corporation Human Annexin A4 (ANXA4) ELISA Kit, 96 tests
Catalog No. 501501820
Encompass_Preferred
The Human Annexin A4 (ANXA4) ELISA Kit allows for quantitative determination of Annexin A4 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.124 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P09525. Cross Activity: No significant cross-reactivity or interference between Annexin A4 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1820 Supplier Biomatik Corporation Supplier No. EKU02431
The Human Annexin A4 (ANXA4) ELISA Kit allows for quantitative determination of Annexin A4 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.124 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P09525. Cross Activity: No significant cross-reactivity or interference between Annexin A4 and analogues was observed. Restriction: For Research Use Only.