Learn More
Biomatik Corporation Human Annexin A6 (ANXA6) ELISA Kit, 96 tests
Catalog No. 501501823
Encompass_Preferred
The Human Annexin A6 (ANXA6) ELISA Kit allows for quantitative determination of Annexin A6 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.056 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08133. Cross Activity: No significant cross-reactivity or interference between Annexin A6 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1823 Supplier Biomatik Corporation Supplier No. EKU02436
The Human Annexin A6 (ANXA6) ELISA Kit allows for quantitative determination of Annexin A6 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.056 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08133. Cross Activity: No significant cross-reactivity or interference between Annexin A6 and analogues was observed. Restriction: For Research Use Only.