Learn More
Biomatik Corporation Human Anti-Immunoglobulin A Antibody (Anti-IgA) ELISA Kit, 96 tests
Catalog No. 501501826
Encompass_Preferred
The Human Anti-Immunoglobulin A Antibody (Anti-IgA) ELISA Kit allows for quantitative determination of Immunoglobulin A in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 6.25-400 ng/mL. Sensitivity: 2.36 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours, 30minutes. Cross Activity: No significant cross-reactivity or interference between Immunoglobulin A and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1826 Supplier Biomatik Corporation Supplier No. EKU02453
The Human Anti-Immunoglobulin A Antibody (Anti-IgA) ELISA Kit allows for quantitative determination of Immunoglobulin A in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 6.25-400 ng/mL. Sensitivity: 2.36 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours, 30minutes. Cross Activity: No significant cross-reactivity or interference between Immunoglobulin A and analogues was observed. Restriction: For Research Use Only.