Learn More
Biomatik Corporation Cattle Anti-Mullerian Hormone (AMH) ELISA Kit, 96 tests
Catalog No. 501501828
Encompass_Preferred
The Cattle Anti-Mullerian Hormone (AMH) ELISA Kit allows for quantitative determination of Anti-Mullerian Hormone in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 493.83-40000 pg/mL. Sensitivity: 177.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P03972. Cross Activity: No significant cross-reactivity or interference between Anti-Mullerian Hormone and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1828 Supplier Biomatik Corporation Supplier No. EKU02458
The Cattle Anti-Mullerian Hormone (AMH) ELISA Kit allows for quantitative determination of Anti-Mullerian Hormone in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 493.83-40000 pg/mL. Sensitivity: 177.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P03972. Cross Activity: No significant cross-reactivity or interference between Anti-Mullerian Hormone and analogues was observed. Restriction: For Research Use Only.