Learn More
Biomatik Corporation Human Anti-Mullerian Hormone (AMH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02459
Encompass_Preferred
The Human Anti-Mullerian Hormone (AMH) ELISA Kit allows for quantitative determination of Anti-Mullerian Hormone in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 185.2-15000 pg/mL. Sensitivity: 83.3 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P03971. Cross Activity: No significant cross-reactivity or interference between Anti-Mullerian Hormone and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1829