Learn More
Biomatik Corporation Human Anti-Myelin Basic Protein Antibody (Anti-MBP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02462
Encompass_Preferred
The Human Anti-Myelin Basic Protein Antibody (Anti-MBP) ELISA Kit allows for quantitative determination of Myelin Basic Protein in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.28 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours, 30minutes. UniProt ID: P02686. Cross Activity: No significant cross-reactivity or interference between Myelin Basic Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1832