Learn More
Biomatik Corporation Human Anti-Myelin Oligodendrocyte Glycoprotein Antibody (Anti-MOG) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02463
Encompass_Preferred
The Human Anti-Myelin Oligodendrocyte Glycoprotein Antibody (Anti-MOG) ELISA Kit allows for quantitative determination of Myelin Oligodendrocyte Glycoprotein in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.21 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours, 30minutes. UniProt ID: Q16653. Cross Activity: No significant cross-reactivity or interference between Myelin Oligodendrocyte Glycoprotein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1833