Learn More
Biomatik Corporation Human Apelin (APLN) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02469
Encompass_Preferred
The Human Apelin (APLN) ELISA Kit allows for quantitative determination of Apelin in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 19.75-1600 pg/mL. Sensitivity: 8.25 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: Q9ULZ1. Cross Activity: No significant cross-reactivity or interference between Apelin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1834