Learn More
Biomatik Corporation Human Apolipoprotein A1 (APOA1) ELISA Kit, 96 tests
Catalog No. 501501837
Encompass_Preferred
The Human Apolipoprotein A1 (APOA1) ELISA Kit allows for quantitative determination of Apolipoprotein A1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 6.25-400 ng/mL. Sensitivity: 2.73 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02647. Cross Activity: No significant cross-reactivity or interference between Apolipoprotein A1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1837 Supplier Biomatik Corporation Supplier No. EKU02480
The Human Apolipoprotein A1 (APOA1) ELISA Kit allows for quantitative determination of Apolipoprotein A1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 6.25-400 ng/mL. Sensitivity: 2.73 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02647. Cross Activity: No significant cross-reactivity or interference between Apolipoprotein A1 and analogues was observed. Restriction: For Research Use Only.