Learn More
Biomatik Corporation Rat Apolipoprotein A1 (APOA1) ELISA Kit, 96 tests
Catalog No. 501501839
Encompass_Preferred
Rat Apolipoprotein A1 (APOA1) ELISA Kit for detecting APOA1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.73ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04639. Gene ID: 25081. Target Synonyms: Apo-A1; ApoA-1 Milano; ProapoA-I; Proapolipoprotein A-I; Truncated apolipoprotein A-I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1839 Supplier Biomatik Corporation Supplier No. EKU02483
Rat Apolipoprotein A1 (APOA1) ELISA Kit for detecting APOA1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.73ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04639. Gene ID: 25081. Target Synonyms: Apo-A1; ApoA-1 Milano; ProapoA-I; Proapolipoprotein A-I; Truncated apolipoprotein A-I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.