Learn More
Biomatik Corporation Human Apolipoprotein A1 Binding Protein (APOA1BP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02486
Encompass_Preferred
The Human Apolipoprotein A1 Binding Protein (APOA1BP) ELISA Kit allows for quantitative determination of Apolipoprotein A1 Binding Protein in Serum, Plasma, Urine, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.061 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q8NCW5. Cross Activity: No significant cross-reactivity or interference between Apolipoprotein A1 Binding Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1840