Learn More
Biomatik Corporation Human Apolipoprotein A2 (APOA2) ELISA Kit, 96 tests
Catalog No. 501501841
Encompass_Preferred
The Human Apolipoprotein A2 (APOA2) ELISA Kit allows for quantitative determination of Apolipoprotein A2 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.69 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02652. Cross Activity: No significant cross-reactivity or interference between Apolipoprotein A2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1841 Supplier Biomatik Corporation Supplier No. EKU02487
The Human Apolipoprotein A2 (APOA2) ELISA Kit allows for quantitative determination of Apolipoprotein A2 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.69 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02652. Cross Activity: No significant cross-reactivity or interference between Apolipoprotein A2 and analogues was observed. Restriction: For Research Use Only.