Learn More
Biomatik Corporation Human Apolipoprotein A4 (APOA4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02491
Encompass_Preferred
The Human Apolipoprotein A4 (APOA4) ELISA Kit allows for quantitative determination of Apolipoprotein A4 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.68 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P06727. Cross Activity: No significant cross-reactivity or interference between Apolipoprotein A4 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1842