Learn More
Biomatik Corporation Mouse Apolipoprotein A4 (APOA4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02492
Encompass_Preferred
Mouse Apolipoprotein A4 (APOA4) ELISA Kit for detecting APOA4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.39-25ng/ml. Sensitivity: 0.17ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06728. Gene ID: 11808. Target Synonyms: ApoA-IV; ApoAIV; Apo-A4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1843