Learn More
Biomatik Corporation Human Apolipoprotein A5 (APOA5) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02495
Encompass_Preferred
The Human Apolipoprotein A5 (APOA5) ELISA Kit allows for quantitative determination of Apolipoprotein A5 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.25-80 ng/mL. Sensitivity: 0.48 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q6Q788. Cross Activity: No significant cross-reactivity or interference between Apolipoprotein A5 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1844