Learn More
Biomatik Corporation Rat Apolipoprotein B (APOB) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02501
Encompass_Preferred
Rat Apolipoprotein B (APOB) ELISA Kit for detecting APOB in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.2-2,000ng/ml. Sensitivity: 12.1ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q7TMA5. Gene ID: 54225. Target Synonyms: Apo-B; FLDB; Apo B-100; Apo B-48. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1847