Learn More
Biomatik Corporation Human Apolipoprotein C3 (APOC3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02511
Encompass_Preferred
The Human Apolipoprotein C3 (APOC3) ELISA Kit allows for quantitative determination of Apolipoprotein C3 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 93.75-6000 pg/mL. Sensitivity: 40.65 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02656. Cross Activity: No significant cross-reactivity or interference between Apolipoprotein C3 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1851