Learn More
Biomatik Corporation Mouse Apolipoprotein C3 (APOC3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02512
Encompass_Preferred
Mouse Apolipoprotein C3 (APOC3) ELISA Kit for detecting APOC3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.37ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P33622. Gene ID: 11814. Target Synonyms: Apo-C3; APOCIII; APOC-III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1852