Learn More
Biomatik Corporation Human Apolipoprotein D (APOD) ELISA Kit, 96 tests
Catalog No. 501501854
Encompass_Preferred
The Human Apolipoprotein D (APOD) ELISA Kit allows for quantitative determination of Apolipoprotein D in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.67 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P05090. Cross Activity: No significant cross-reactivity or interference between Apolipoprotein D and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1854 Supplier Biomatik Corporation Supplier No. EKU02515
The Human Apolipoprotein D (APOD) ELISA Kit allows for quantitative determination of Apolipoprotein D in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.67 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P05090. Cross Activity: No significant cross-reactivity or interference between Apolipoprotein D and analogues was observed. Restriction: For Research Use Only.