Learn More
Biomatik Corporation Human Apolipoprotein E (APOE) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02517
Encompass_Preferred
The Human Apolipoprotein E (APOE) ELISA Kit allows for quantitative determination of Apolipoprotein E in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 4.7-300 ng/mL. Sensitivity: 1.9 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02649. Cross Activity: No significant cross-reactivity or interference between Apolipoprotein E and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1855